Dynabeads™ Protein G IP Kit and Magnet Starter Pack
Invitrogen™
Dynabeads™ Protein G IP Kit and Magnet Starter Pack
The Dynabeads Protein G IP Kit and Magnet Starter Pack combines Dynabeads Protein G magnetic beads for immunoprecipitation (including theRead more
Have Questions?
Catalog NumberQuantity
10019D40 reactions
Catalog number 10019D
Price (INR)
76,422.00
Online offer
Ends: 31-Dec-2026
113,217.00
Save 36,795.00 (32%)
Each
Quantity:
40 reactions
Price (INR)
76,422.00
Online offer
Ends: 31-Dec-2026
113,217.00
Save 36,795.00 (32%)
Each
The Dynabeads Protein G IP Kit and Magnet Starter Pack combines Dynabeads Protein G magnetic beads for immunoprecipitation (including the required buffers) and the magnet preferred for use with 1.5 mL microcentrifuge tubes for purchasing convenience. The beads and buffers are sufficient for 40 reactions, and the magnet holds 16 x 1.5 mL microcentrifuge tubes. The Immunoprecipitation Kit - Dynabeads Protein G with buffers, stand-alone Dynabeads Protein G beads, and the DynaMag-2 magnet are also available separately.
The product contains:
1 x Immunoprecipitation Kit - Dynabeads Protein G*
1 x DynaMag-2 Magnet
*The kit contains 2 mL Dynabeads Protein G beads, 16 mL Antibody Binding & Washing Buffer, 28 mL Washing Buffer, and 1 mL Elution Buffer.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Application)Immunoprecipitation
Kit Contents2 mL Dynabeads Protein G beads; 16 mL Antibody Binding and Washing Buffer; 28 mL Washing Buffer; 1 mL Elution Buffer; 1 x DynaMag-2 Magnet
Ligand TypeProtein G
Quantity40 reactions
Shipping ConditionRoom Temperature
Product LineDynaMag, Dynabeads
TypeIP Kit and Magnet Starter Pack
Unit SizeEach
Contents & Storage
Store Dynabeads at 2–8°C.