CEDIA™ Buprenorphine Drugs of Abuse Calibrators
Actual product may vary
CEDIA™ Buprenorphine Drugs of Abuse Calibrators
CEDIA™ Buprenorphine Drugs of Abuse Calibrators
Thermo Scientific™

CEDIA™ Buprenorphine Drugs of Abuse Calibrators

Utilize the globally valued accuracy, precision and lot-to-lot dependability of CEDIA™ Buprenorphine Drugs of Abuse Calibrators.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantityConcentration
1002441 x 5 mL50 ng/mL
1002411 x 7.5 mL0 ng/nL
1002421 x 5 mL5 ng/mL
1002431 x 5 mL20 ng/mL
1002451 x 5 mL75 ng/mL
Catalog number 100244
Price (INR)
-
Quantity:
1 x 5 mL
Concentration:
50 ng/mL
Utilize the globally valued accuracy, precision and lot-to-lot dependability of CEDIA™ Buprenorphine Drugs of Abuse Calibrators, which are optimized for an array of clinical chemistry analyzers. The drugs of abuse testing scope is expanding rapidly, and CEDIA assays meet this challenge by offering one of the most extensive menus for drugs of abuse testing on the market.
  • Qualitative or semi-quantitative options for results
  • Excellent correlation to GC/MS
  • Applications for an array of clinical chemistry analyzers
  • Variety of calibrators and multiconstituent control sets
For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  
Specifications
Concentration50 ng/mL
DescriptionCEDIA; 50ng/mL; 5mL
Packaging1 Bottle
Quantity1 x 5 mL
TypeCalibrator
Detectable AnalytesBuprenorphine
Sample TypeUrine
Unit SizeEach
Contents & Storage
2°C to 8°C