CEDIA™ Buprenorphine Drugs of Abuse Control Set
Product Image
Thermo Scientific™

CEDIA™ Buprenorphine Drugs of Abuse Control Set

Get world-renowned accuracy, dependability and stability with the Thermo Scientific™ CEDIA™ Buprenorphine Drugs of Abuse Control Set, which is optimized for most analyzers.
Have Questions?
Catalog NumberQuantity
1002462 x 5 mL
Catalog number 100246
Price (INR)
-
Request A Quote
Quantity:
2 x 5 mL
Get world-renowned accuracy, dependability and stability with the Thermo Scientific™ CEDIA™ Buprenorphine Drugs of Abuse Control Set, which is optimized for most analyzers. CEDIA assays offer one of the most extensive menus on the market for drugs of abuse testing.
  • Qualitative or semi-quantitative options for results
  • Excellent correlation to GC/MS
  • Applications for an array of clinical chemistry analyzers
  • Variety of calibrators and multiconstituent control sets
For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  
Specifications
Concentration5 ng/mL, Low (3ng/mL), High (7ng/mL)
DescriptionCEDIA Buprenorphine Control Set: Low 3 and High 7
LevelHigh, Low
Quantity2 x 5 mL
TypeControl
Detectable AnalytesBuprenorphine
Sample TypeUrine
Unit SizeEach