Product Image
Thermo Scientific™

SickleScreen™ Control Set, 4 x 0.5mL

Control Kit for use with Thermo Scientific SickleScreen Assay Set, 22-251050
Have Questions?
Catalog NumberQuantity
100251TS
also known as 100251
4 vials x 0.5mL bottles
Catalog number 100251TS
also known as 100251
Price (INR)
-
Quantity:
4 vials x 0.5mL bottles
For use with Thermo Scientific SickleScreen Assay Set.
For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  
Specifications
DescriptionSickleScreen Control Set
IncludesPositive and Negative Controls; Each level -4 bottles each level x 5 ml fill, Reconstitutiom Control Kit - 2 bottles x 4 mL per vial
Quantity4 vials x 0.5mL bottles
Unit SizeEach