DRI™ Oxycodone Drugs of Abuse Calibrators
DRI™ Oxycodone Drugs of Abuse Calibrators
Actual product may vary
Thermo Scientific™

DRI™ Oxycodone Drugs of Abuse Calibrators

Utilize the globally valued accuracy, precision and lot-to-lot dependability of Thermo Scientific™ DRI™ Oxycodone Drugs of Abuse Calibrators.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberConcentration
100252500 ng/mL
1002531000 ng/mL
100251TS1
also known as 100251
300 ng/mL
100250TS1
also known as 100250
100 ng/mL
Catalog number 100252
Price (INR)
-
Request A Quote
Concentration:
500 ng/mL

Utilize the globally valued accuracy, precision and lot-to-lot dependability of Thermo Scientific™ DRI™ Oxycodone Drugs of Abuse Calibrators, which are optimized for an array of clinical chemistry analyzers. The drugs of abuse testing scope is expanding rapidly, and DRI assays meet this challenge by offering one of the most extensive menus for drugs of abuse testing on the market.

  • Qualitative or semi-quantitative options for results
  • Excellent correlation to GC/MS
  • Applications for an array of clinical chemistry analyzers
  • Variety of calibrators and multiconstituent control sets
For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  
Specifications
Concentration500 ng/mL
DescriptionDRI; 500ng/mL; 10mL
Packaging1 Bottle
Quantity1 x 10 mL
TypeCalibrator
Detectable AnalytesOxycodone
Sample TypeUrine
Unit SizeEach
Contents & Storage
2°C to 8°C