DRI™ Creatinine-Detect™ Specimen Validity Test Controls
Product Image
Thermo Scientific™
DRI™ Creatinine-Detect™ Specimen Validity Test Controls
Combat the use of additives and adulterants to mask controlled substances with Thermo Scientific™ DRI™ Creatinine-Detect™ Specimen Validity Test Controls.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberConcentration
100273TS
also known as 100273
1.3 mg/dL
100274TS
also known as 100274
7.5 mg/dL
100275TS
also known as 100275
23 mg/dL
Catalog number 100273TS
also known as 100273
Price (INR)
-
Request A Quote
Concentration:
1.3 mg/dL
Combat the use of additives and adulterants to mask controlled substances in urine samples with Thermo Scientific™ DRI™ Creatinine-Detect™ Specimen Validity Test Controls. Liquid and ready-to-use, these controls can be performed on a variety of general chemistry analyzers. As new adulterant trends sweep through the testing pool, rely on our fast-track product development team to deliver a timely, economical detection solution.
  • One of the most extensive menus of tests available
  • Associated calibrators and controls offered in convenient sizes
For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  
Specifications
Concentration1.3 mg/dL
DescriptionDRI™ Creatinine Specimen Validity Test Controls
Quantity1 x 25 mL
TypeControl
Detectable AnalytesCreatinine
Sample TypeUrine
Unit SizeEach
Contents & Storage
2°C to 8°C