Pacific Hemostasis™ Activated Partial Thromboplastin Time (APTT) Reagents
Actual product may vary
Thermo Scientific™

Pacific Hemostasis™ Activated Partial Thromboplastin Time (APTT) Reagents

Fisherbrand™ Pacific Hemostasis Activated Partial Thromboplastin Time (aPTT) Reagents are manufactured with up to two years extended shelf life.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescriptionQuantity
100312TS
also known as 100312
KONTACT (Micronized Silica Activator)10 x 10 mL
100309CaCl2, 0.025M10 x 4 mL
100304TS
also known as 100304
CaCl2, 0.02M10 x 10 mL
100314TS
also known as 100314
CaCl2, 0.025M10 x 10 mL
100402TS
also known as 100402
APTT-XL (Ellagic Acid Activator)10 x 4 mL
100403aPTT-XL10 x 10 mL
100308KONTACT (Particulate Activator)10 x 4 mL
Catalog number 100312TS
also known as 100312
Price (INR)
-
Description:
KONTACT (Micronized Silica Activator)
Quantity:
10 x 10 mL
Identify acquired or congenital disorders to the intrinsic and common pathways with Thermo Scientific™ Pacific Hemostasis™ Activated Partial Thromboplastin Time (APTT) Reagents. Widely accepted as a means of monitoring patients on oral anticoagulant therapy such as Heparin, reagents are manufactured with up to two years extended shelf life and have excellent lot-to-lot consistency.
Features:
  • 10mL vials can complete ∽1000 full volume, single determinations; 4mL vials can complete ∽400 full volume, single determinations
  • Open vial stability: 30 days at 2° to 8°C

aPTT-XL (Ellagic Acid Activator) (Product No. 10-0402):

  • Superior factor sensitivity and responsiveness
  • Increased sensitivity to heparin and lupus-like inhibitors

KONTACT (Particulate Activator) (Product No. 01-0402):

  • Higher sensitivity to Factor VIII
  • Average Normal Values: 26 - 33 Seconds
For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  
Specifications
DescriptionKONTACT (Micronized Silica Activator)
Quantity10 x 10 mL
Shelf LifeUp to 2 Years
TypeActivated Partial Thromboplastin Time (APTT)
Unit SizeEach