Pacific Hemostasis™ Prothrombin Time (PT) Reagents
Actual product may vary
Thermo Scientific™

Pacific Hemostasis™ Prothrombin Time (PT) Reagents

Detect acquired or congenital disorders affecting the extrinsic and common pathways.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantityType
100354TS
also known as 100354
10 x 4 mLThromboplastin-DS
10036210 x 10 mLThromboplastin-DS
100356TS
also known as 100356
10 x 4 mLProthrombin Time (PT)
10035710 x 10 mLProthrombin Time (PT)
Catalog number 100354TS
also known as 100354
Price (INR)
-
Quantity:
10 x 4 mL
Type:
Thromboplastin-DS
Detect acquired or congenital disorders affecting the extrinsic and common pathways with Thermo Scientific™ Pacific Hemostasis™ Prothrombin Time (PT) Reagents. Widely accepted as a means of monitoring patients on oral anticoagulant therapy such as Warfarin/Coumadin, reagents are manufactured with up to three years extended shelf life and have excellent lot-to-lot consistency.
Features:
  • Compatible with automated, semi-automated or manual systems
  • Reconstitution stability: 7 days at 2° to 8°C (36° to 46°F)
  • Dried rabbit brain thromboplastin

Thromboplastin-D with CaCl2 (Product No. 10-0356, 10-0357):

  • Target ISI value of 1.7 to 2.2
  • Expected normal values of 12 to 15 seconds

Thromboplastin-DS (Product No. 10-0362, 10-0354):

  • Target ISI value of 1.0 to 1.3
  • Expected normal values of 12 to 15 seconds
For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  
Specifications
Quantity10 x 4 mL
Shelf LifeUp to 3 Years
TypeThromboplastin-DS
Unit SizeEach