Pacific Hemostasis™ Imidazole Buffered Saline
Actual product may vary
Thermo Scientific™

Pacific Hemostasis™ Imidazole Buffered Saline

Fisherbrand™ Pacific Hemostasis Fibrinogen/Fibrin Analysis is a complete line of assays used to screen and quantify plasma fibrinogen based on the Clauss method.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberTypeQuantity
100600TS
also known as 100600
Fibrinogen Assay100 Determinations
100606TS
also known as 100606
Bovine Thrombin10 x 5 mL
100602TS
also known as 100602
Fibrinogen Reference10 x 1 mL
100011Thrombin Time Reagent10 x 1 mL
100601TS
also known as 100601
Bovine Thrombin10 x 2 mL
100647TS
also known as 100647
Imidazole Buffered Saline2 x 135 mL
Catalog number 100600TS
also known as 100600
Price (INR)
-
Type:
Fibrinogen Assay
Quantity:
100 Determinations
Screen and quantify plasma fibrinogen based on the Clauss method with Thermo Scientific™ Pacific Hemostasis™ Fibrinogen/Fibrin Analysis.
Fibrinogen Assay Set:
  • Quantifies plasma fibrinogen levels
  • Detects acquired and congenital hemorrhagic disorders
  • Components available separately
Thrombin Time Assay:
  • Provides qualitative determination
  • Useful in screening for fibrinogen disorders by measuring the formation of the fibrin clot
  • Reconstituted stability: 8 hours at 2° to 8°C

Ordering Information:

Products can be purchased as a set or individually.

For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  
Specifications
Quantity100 Determinations
TypeFibrinogen Assay
Unit SizeEach
Contents & Storage
2°C to 8°C