Pacific Hemostasis™ Imidazole Buffered Saline
Pacific Hemostasis™ Imidazole Buffered Saline
Actual product may vary
Thermo Scientific™

Pacific Hemostasis™ Imidazole Buffered Saline

Fisherbrand™ Pacific Hemostasis Fibrinogen/Fibrin Analysis is a complete line of assays used to screen and quantify plasma fibrinogen based on the Clauss method.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberTypeQuantity
100606TS
also known as 100606
Bovine Thrombin10 x 5 mL
100602TS
also known as 100602
Fibrinogen Reference10 x 1 mL
100011Thrombin Time Reagent10 x 1 mL
100600TS
also known as 100600
Fibrinogen Assay100 Determinations
100601TS
also known as 100601
Bovine Thrombin10 x 2 mL
100647TS
also known as 100647
Imidazole Buffered Saline2 x 135 mL
Catalog number 100606TS
also known as 100606
Price (INR)
-
Request A Quote
Type:
Bovine Thrombin
Quantity:
10 x 5 mL
Screen and quantify plasma fibrinogen based on the Clauss method with Thermo Scientific™ Pacific Hemostasis™ Fibrinogen/Fibrin Analysis.
Fibrinogen Assay Set:
  • Quantifies plasma fibrinogen levels
  • Detects acquired and congenital hemorrhagic disorders
  • Components available separately
Thrombin Time Assay:
  • Provides qualitative determination
  • Useful in screening for fibrinogen disorders by measuring the formation of the fibrin clot
  • Reconstituted stability: 8 hours at 2° to 8°C

Ordering Information:

Products can be purchased as a set or individually.

Specifications
Quantity10 x 5 mL
TypeBovine Thrombin
Unit SizeEach
Contents & Storage
2°C to 8°C