Pacific Hemostasis™ FDP Assay
Actual product may vary
Thermo Scientific™
Pacific Hemostasis™ FDP Assay
Fisherbrand™ Pacific Hemostasis FDP Assay is a fast, easy-to-use latex agglutination assay for use in measuring fibrinogen degradation products (FDP) in serum and urine.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
1006511 x 5 mL
1006652 x 100 mL
100650TS
also known as 100650
30 Determinations
1006563 x 2 mL
10065430 Tubes
Catalog number 100651
Price (INR)
-
Quantity:
1 x 5 mL
Measure fibrinogen degradation products (FDP) in serum and urine with Thermo Scientific™ Pacific Hemostasis™ FDP Assay, a fast, easy-to-use latex agglutination assay used in the diagnosis of disseminated intravascular coagulation (DIC), deep vein thrombosis (DVT), pulmonary embolism (PE), certain pregnancy disorders and myocardial infarction.
Features:
  • Latex sensitivity of 1ug/mL of FDP
  • No pretreatment of heparin specimen
  • No repeat testing due to false positives from heparin interference
For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  
Specifications
DescriptionLatex Reagent
Quantity1 x 5 mL
Unit SizeEach