Pacific Hemostasis™ Factor Assays
Actual product may vary
Thermo Scientific™
Pacific Hemostasis™ Factor Assays
Fisherbrand™ Pacific Hemostasis Factor Assays are used for the detection and quantification of factor deficiencies in patient plasmas.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberType
100800Factor VIII Deficient Plasma
100050TS1
also known as 100050
Factor V Deficient Plasma
100644TS
also known as 100644
Barbitol Buffered Saline
Catalog number 100800
Price (INR)
-
Type:
Factor VIII Deficient Plasma
Detect and quantify factor deficiencies in patient plasmas with Thermo Scientific™ Pacific Hemostasis™ Factor Assays.
Pacific Hemostasis Factor Assays include a complete line of lyophilized congenital factor deficient plasmas for performing factor assays.
  • All factor deficient plasmas are prepared from congenitally deficient patient donors or immunodepleted plasma, with the exception of Factor II.
  • Factor II Deficient Plasma is a mix of human and bovine plasma.
  • Plasmas are packaged 10 × 1mL; Diluting Fluid is packaged 2 × 100mL.
For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  
Specifications
TypeFactor VIII Deficient Plasma
Unit SizeEach