Max-Bev CO<sub>2</sub> Purity Monitoring System
Thermo Scientific™
Max-Bev CO2 Purity Monitoring System
The Thermo Scientific™ MAX-Bev™ CO₂ Purity Monitoring System is a reliable analyzer for monitoring CO₂ purity and detecting trace impurities at low levels (up to 100% +/- 0.02%). It provides sensitive, accurate measurements, meeting regulatory standards. The system supports industrial applications like beverage production quality assurance and CO₂ stream monitoring in pipelines, crucial for Carbon Capture Utilization and Storage (CCUS) processes.
Have Questions?
Catalog NumberSample Type
101-0411CO2 Purity
Catalog number 101-0411
Price (INR)
-
Sample Type:
CO2 Purity
MAX-Bev Gas Analyzer product overview
Engineered for demanding industrial environments, the MAX-Bev CO₂ Purity Monitoring System delivers real-time, continuous monitoring of carbon dioxide levels, ensuring that even the smallest impurities are detected. This capability is essential for maintaining the purity of CO₂ streams, which is vital for both beverage QA and for various industrial processes like CCUS.
Beverage QA: CO₂ analysis helps achieve customer safety, satisfaction, and brand loyalty
The MAX-Bev System is a fully automated system designed to analyze carbonation streams for all gaseous impurities using state-of-the-art FTIR technology. It is capable of simultaneous accurate measurement of CO₂ absolute purity and more than 20 impurities down to ppb levels, as specified by the International Society of Beverage Technologists (ISBT) and the European Industrial Gases Association (EIGA).
The elegant design permits thorough monitoring of the input process gas streams throughout all points of the carbonation process (delivery tanker, pre- and post- filtration), along with zero gas and validation gas. Learn how our CO₂ purity monitoring solutions help meet rigorous regulatory standards for beverage-grade certification.
Max-Bev Analyzer features and benefits for beverage analysis:
  • Measures to levels that easily meet and exceed ISBT and EIGA requirements
  • Incorporates highly automated, user-friendly software – pushbutton operation, no technical expertise required
  • Provides reliable “hands-off” operation using proven FTIR technology
  • Has low cost of ownership with easy maintenance
  • Saves operator time by replacing unreliable and cumbersome UV-fluorescence and wet chemistry techniques
  • Offers total software control, maintained via the large front panel touchscreen or keyboard.
  • Uses common communication protocols for data logging, making for easy incorporation into existing plant designs.
  • Can easily generate and print Certificates of Analysis
CCUS: Protect and maintain valuable pipeline infrastructure
The MAX-Bev System is emerging as a vital tool for CCUS applications, essential for safeguarding pipeline infrastructure. The system utilizes FTIR spectroscopy for real-time measurement of low-level (ppm or even ppb) impurities within the captured CO₂. With our instrument, industry professionals can monitor purity and analyze the change in impurity levels, helping to ensure the integrity of the carbon dioxide pipeline and preventing the formation of acidic conditions. The MAX-Bev System is a factory-ready solution; it is pre-calibrated and can be used for CO₂ certification directly after installation. Explore how the MAX-Bev System can facilitate the shift to a zero-carbon future.
Max-Bev Analyzer features and benefits for pipeline infrastructure:
  • Measures CO₂ absolute purity to 100±0.02% to ensure carbon capture stream quality
  • Utilizes workflows to achieve product quality specifications automatically
  • Can provide continuous, on-line analysis of absolute purity and impurities
  • Performs automatic gas sampling and analysis
  • Possesses the connectivity needed for reporting and data publishing
  • Can customize list of gases to analyze depending on sample matrix
  • Has customizable alarm notifications for hardware and gas concentrations
  • Requires no user intervention to calibrate, thus minimizing maintenance and downtime
Specifications
Sample TypeCO2 Purity
Voltage208/240 V
Unit SizeEach