Product Image
Have Questions?
Catalog number 10100
Price (INR)
-
Frequently asked questions (FAQs)
How long can serum be stored at 2-8 degrees C?
Serum can be kept at 2-8 degrees C for up to 4 weeks.
Find additional tips, troubleshooting help, and resources within ourMammalian Cell Culture Basics Support Center.