Rec Protein A-HRP
Invitrogen™
Rec Protein A-HRP
This recombinant protein A is purified from cytosol of an E. coli strain expressing the genes coding for Staphylococcus aureusRead more
Have Questions?
Catalog NumberQuantity
1011233 mg
Catalog number 101123
Price (INR)
17,640.00
Online offer
Ends: 30-Sep-2026
29,399.00
Save 11,759.00 (40%)
Each
Quantity:
3 mg
Price (INR)
17,640.00
Online offer
Ends: 30-Sep-2026
29,399.00
Save 11,759.00 (40%)
Each
This recombinant protein A is purified from cytosol of an E. coli strain expressing the genes coding for Staphylococcus aureus protein A. This version is conjugated with highly purified horseradish peroxidase.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
FormSolution
Protein SubtypeProtein A
Shipping ConditionWet Ice
ConjugateHRP
Quantity3 mg
Unit SizeEach
Contents & Storage
2-8°C. Do Not Freeze.