2-Aminoethyl hydrogen sulfate, 99%
2-Aminoethyl hydrogen sulfate, 99%
2-Aminoethyl hydrogen sulfate, 99%
Acros Organics

2-Aminoethyl hydrogen sulfate, 99%

CAS: 926-39-6 | C2H7NO4S | 141.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
2.5 kg
Packaging:
View all
Plastic bottle
Catalog number 103645000
also known as 10364
HSN 29221990
Price (INR)
64,765.00
Each
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (INR)
64,765.00
Each
Chemical Identifiers
CAS926-39-6
IUPAC Name(2-aminoethoxy)sulfonic acid
Molecular FormulaC2H7NO4S
InChI KeyWSYUEVRAMDSJKL-UHFFFAOYSA-N
SMILESNCCOS(O)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)White to light beige
Appearance (Form)Crystalline powder
Titration with NaOH>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only