2-Amino-2-methyl-1-propanol, 99%
2-Amino-2-methyl-1-propanol, 99%
2-Amino-2-methyl-1-propanol, 99%
Acros Organics

2-Amino-2-methyl-1-propanol, 99%

CAS: 124-68-5 | C4H11NO | 89.14 g/mol
Have Questions?
Catalog NumberQuantityPackaging
104060010
also known as 10406
1 LGlass bottle
Catalog number 104060010
also known as 10406
HSN 29221990
Price (INR)
13,458.00
Each
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
13,458.00
Each
Chemical Identifiers
CAS124-68-5
IUPAC Name2-amino-2-methylpropan-1-ol
Molecular FormulaC4H11NO
InChI KeyCBTVGIZVANVGBH-UHFFFAOYSA-N
SMILESCC(C)(N)CO
View more
SpecificationsSpecification SheetSpecification Sheet
Water=<0.8 %
Appearance (Form)Low melting solid
Infrared spectrumConforms
Appearance (Color)Colorless
Titration with HClO4>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only