2-Aminonicotinic acid, 98%
2-Aminonicotinic acid, 98%
2-Aminonicotinic acid, 98%
Acros Organics

2-Aminonicotinic acid, 98%

CAS: 5345-47-1 | C6H6N2O2 | 138.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 104160050
also known as 10416
HSN 29333990
Price (INR)
5,066.00
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
5,066.00
Each
Chemical Identifiers
CAS5345-47-1
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Off-white to light yellow
Infrared spectrumConforms
HPLC>=97.5 %
Appearance (Form)Crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only