2-Amino-6-picoline, 98%
2-Amino-6-picoline, 98%
2-Amino-6-picoline, 98%
Acros Organics

2-Amino-6-picoline, 98%

CAS: 1824-81-3 | C6H8N2 | 108.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
500 g
2.5 kg
Packaging:
Glass bottle
Catalog number 104401000
also known as 10440
HSN 29333990
Price (INR)
5,326.00
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
5,326.00
Each
Chemical Identifiers
CAS1824-81-3
IUPAC Name6-methylpyridin-2-amine
Molecular FormulaC6H8N2
InChI KeyQUXLCYFNVNNRBE-UHFFFAOYSA-N
SMILESCC1=CC=CC(N)=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to white to yellow to light orange
Appearance (Form)Low melting crystalline solid
Infrared spectrumConforms
Melting point40°C to 45°C
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only