p-Anisaldehyde, 99+%
p-Anisaldehyde, 99+%
p-Anisaldehyde, 99+%
Acros Organics

p-Anisaldehyde, 99+%

CAS: 123-11-5 | C8H8O2 | 136.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
5 mL
500 mL
2.5 L
Packaging:
Glass bottle
Catalog number 104801000
HSN 29124999
Price (INR)
7,314.00
Each
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
7,314.00
Each
Chemical Identifiers
CAS123-11-5
IUPAC Name4-methoxybenzaldehyde
Molecular FormulaC8H8O2
InChI KeyZRSNZINYAWTAHE-UHFFFAOYSA-N
SMILESCOC1=CC=C(C=O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=99.0 %
Appearance (Form)Liquid
Relative density(D20/4°C) 1.120 to 1.124
Appearance (Color)Clear colorless to yellow
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only