Anthracene, 99%
Anthracene, 99%
Anthracene, 99%
Acros Organics

Anthracene, 99%

CAS: 120-12-7 | C14H10 | 178.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
500 g
2.5 kg
Packaging:
View all
Plastic bottle
Catalog number 104861000
HSN 29029090
Price (INR)
8,143.00
Each
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (INR)
8,143.00
Each
Chemical Identifiers
CAS120-12-7
IUPAC Nameanthracene
Molecular FormulaC14H10
InChI KeyMWPLVEDNUUSJAV-UHFFFAOYSA-N
SMILESC1=CC2=CC3=CC=CC=C3C=C2C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow to beige to green-beige
Appearance (Form)Powder or crystals
Infrared spectrumConforms
Melting point212°C to 219°C
GC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only