DL-Aspartic acid, 99+%
DL-Aspartic acid, 99+%
DL-Aspartic acid, 99+%
Acros Organics

DL-Aspartic acid, 99+%

CAS: 617-45-8 | C4H7NO4 | 133.10 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
5 g
1 kg
Packaging:
View all
Plastic Bottle
Catalog number 105032500
also known as 10503
HSN 29224990
Price (INR)
7,267.00
Each
Quantity:
250 g
Packaging:
Plastic Bottle
Request bulk or custom format
Chemical Identifiers
CAS617-45-8
IUPAC Name2-aminobutanedioic acid
Molecular FormulaC4H7NO4
InChI KeyCKLJMWTZIZZHCS-UHFFFAOYNA-N
SMILESNC(CC(O)=O)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow
Appearance (Form)Powder or crystals
Infrared spectrumConforms
Titration with NaOH99.0 to 101.0 %
Heavy metals (as Pb)=<10 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only