Benzoic anhydride, 98%
Benzoic anhydride, 98%
Benzoic anhydride, 98%
Acros Organics

Benzoic anhydride, 98%

CAS: 93-97-0 | C14H10O3 | 226.23 g/mol
Have Questions?
Catalog NumberQuantityPackaging
105500025
also known as 10550
2.5 kgPlastic bottle
Catalog number 105500025
also known as 10550
HSN 29089990
Price (INR)
69,665.00
Each
Quantity:
2.5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (INR)
69,665.00
Each
Chemical Identifiers
CAS93-97-0
IUPAC Namebenzoyl benzoate
Molecular FormulaC14H10O3
InChI KeyCHIHQLCVLOXUJW-UHFFFAOYSA-N
SMILESO=C(OC(=O)C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
HPLC>=97.5 %
Melting point38°C to 45°C
Appearance (Form)Crystalline powder or crystals
Appearance (Color)White to cream to light beige
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only