Benzophenone hydrazone, 98+%
Benzophenone hydrazone, 98+%
Benzophenone hydrazone, 98+%
Acros Organics

Benzophenone hydrazone, 98+%

CAS: 5350-57-2 | C13H12N2 | 196.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
500 g
Packaging:
Glass bottle
Catalog number 105571000
also known as 10557
HSN 29280090
Price (INR)
4,687.00
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
4,687.00
Each
Chemical Identifiers
CAS5350-57-2
IUPAC Name(diphenylmethylidene)hydrazine
Molecular FormulaC13H12N2
InChI KeyQYCSNMDOZNUZIT-UHFFFAOYSA-N
SMILESNN=C(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
GC>=98.0 %
Water=<0.5 %
Appearance (Color)White to yellow
Appearance (Form)Crystalline powder or needles
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only