PCR SuperMix is a ready-to-use mixture of DNA polymerase, salts, magnesium, and dNTPs for efficient PCR amplification.
Have Questions?
Change view
Catalog Number
No. of Reactions
10572114
100 Reactions
10572152
500 Reactions
10572163
5000 Reactions
3 Options
Catalog number 10572114
Price (INR)
3,640.00
Online offer
Ends: 31-Mar-2026
5,392.00
Save 1,752.00 (32%)
Each
Add to cart
No. of Reactions:
100 Reactions
Price (INR)
3,640.00
Online offer
Ends: 31-Mar-2026
5,392.00
Save 1,752.00 (32%)
Each
Add to cart
PCR SuperMix is a ready-to-use mixture of DNA polymerase, salts, magnesium, and dNTPs for efficient PCR amplification. All you have to do is add template and primers, reducing set-up time by half (Figure 1).
PCR SuperMix contains recombinant Taq DNA polymerase for routine PCR of fragments up to 5 kb.
For Research Use Only. Not for use in diagnostic procedures.