Product Image
Invitrogen™

PCR Supermix (Brasil)

PCR SuperMix is a ready-to-use mixture of DNA polymerase, salts, magnesium, and dNTPs for efficient PCR amplification.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberNo. of Reactions
10572152500 Reactions
10572114100 Reactions
105721635000 Reactions
Catalog number 10572152
Price (INR)
-
No. of Reactions:
500 Reactions
PCR SuperMix is a ready-to-use mixture of DNA polymerase, salts, magnesium, and dNTPs for efficient PCR amplification. All you have to do is add template and primers, reducing set-up time by half (Figure 1).

PCR SuperMix contains recombinant Taq DNA polymerase for routine PCR of fragments up to 5 kb.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Fidelity (vs. Taq)1X
FormatTube
Hot StartNo
No. of Reactions500 Reactions
Overhang3'-A
PolymeraseTaq DNA Polymerase
Product TypePCR SuperMix
Quantity500 reactions
Reaction FormatSuperMix or Master Mix
Shipping ConditionDry Ice
Size (Final Product)5 kb or less
Starting MaterialDNA
Concentration1.1X
For Use With (Application)Standard PCR
GC-Rich PCR PerformanceLow
Reaction SpeedStandard
Unit SizeEach
Contents & Storage
• PCR Supermix, 4 x 5.625 mL

Store at -20°C in a non-frost-free freezer.