Benzoylformic acid, 97%
Benzoylformic acid, 97%
Benzoylformic acid, 97%
Benzoylformic acid, 97%
Acros Organics

Benzoylformic acid, 97%

CAS: 611-73-4 | C8H6O3 | 150.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Packaging:
Glass bottle
Catalog number 105761000
also known as 10576
HSN 29183090
Price (INR)
42,656.00
Each
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
42,656.00
Each
Add to cart
Chemical Identifiers
CAS611-73-4
IUPAC Name2-oxo-2-phenylacetic acid
Molecular FormulaC8H6O3
InChI KeyFAQJJMHZNSSFSM-UHFFFAOYSA-N
SMILESOC(=O)C(=O)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Melting point60°C to 69°C
Appearance (Color)White to light yellow
Titration with NaOH>=96.0 %
Appearance (Form)Crystalline powder or crystals and/or chunks
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only