Benzylamine, 99%, pure
Benzylamine, 99%, pure
Benzylamine, 99%, pure
Acros Organics

Benzylamine, 99%, pure

CAS: 100-46-9 | C7H9N | 107.15 g/mol
Have Questions?
Catalog NumberQuantityPackaging
1058500252.5 LGlass bottle
Catalog number 105850025
HSN 29214990
Price (INR)
27,719.00
Each
Quantity:
2.5 L
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
27,719.00
Each
Chemical Identifiers
CAS100-46-9
IUPAC Name1-phenylmethanamine
Molecular FormulaC7H9N
InChI KeyWGQKYBSKWIADBV-UHFFFAOYSA-N
SMILESNCC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
Refractive index1.5420 to 1.5450 (20°C, 589 nm)
GC>=98.5 %
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only