O-Benzylhydroxylamine Hydrochloride, 99%
O-Benzylhydroxylamine Hydrochloride, 99%
O-Benzylhydroxylamine Hydrochloride, 99%
Acros Organics

O-Benzylhydroxylamine Hydrochloride, 99%

CAS: 2687-43-6 | C7H9NO·HCl | 159.62 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Packaging:
Glass bottle
Catalog number 105971000
also known as 10597
HSN 29225090
Price (INR)
41,070.00
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
41,070.00
Each
Chemical Identifiers
CAS2687-43-6
IUPAC Namehydrogen O-benzylhydroxylamine chloride
Molecular FormulaC7H10ClNO
InChI KeyHYDZPXNVHXJHBG-UHFFFAOYSA-N
SMILES[H+].[Cl-].NOCC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or flakes
Infrared spectrumConforms
Titration Argentometric>=98.5 %
Appearance (Color)White to off-white
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only