Dionex™ EG Eluent Generator Kits
Dionex™ EG Eluent Generator Kits
Thermo Scientific™
Dionex™ EG Eluent Generator Kits
Run ion chromatography gradients as easily as isocratic by installing the Thermo Scientific™ Dionex™ EG Cartridge Kits in the Dionex ICS-6000 EG Eluent Generator.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescriptionFor Use With (Equipment)
106-60001RFIC Eluent DegasserDionex Inuvion IC System
075522Analytical EG KitDionex ICS-6000 EG Eluent Generator
074459Capillary EG Cartridge KitDionex ICS-5000+ IC System (Capillary)
22181-60202Capillary EG KitDionex ICS-6000 EG Eluent Generator
22181-60951Dionex RFIC+ Dual EGC Degasser Dionex ICS-6000 HPIC System in Dual EGC Mode
22181-62038RFIC+ Dual EGC Degasser Ship KitDionex ICS-6000 HPIC System in Dual EGC Mode
22181-62048RFIC+ Dual EGC Vacuum Degas KitDionex ICS-6000 HPIC System in Dual EGC Mode
Catalog number 106-60001
Price (INR)
-
Request A Quote
Description:
RFIC Eluent Degasser
For Use With (Equipment):
Dionex Inuvion IC System
Achieve reproducible and reliable chromatography separations using Thermo Scientific™ Dionex™ Reagent-Free IC (RFIC™) technologies. Dionex Eluent Generator accessories allow you to operate your RFIC system with minimal effort through plug-and-play setup.
Operate your ion chromatography system with minimal effort when using Dionex EG Eluent Generator accessories.
Specifications
DescriptionRFIC Eluent Degasser
Flow Rate0.1 to 3.00 mL/min
For Use With (Equipment)Dionex Inuvion IC System
Pressure5000 psi
QuantityEach
TypeAnalytical Degasser Module
Unit SizeEach