N-Methylbenzylamine, 97%
N-Methylbenzylamine, 97%
N-Methylbenzylamine, 97%
Acros Organics

N-Methylbenzylamine, 97%

CAS: 103-67-3 | C8H11N | 121.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 mL
500 mL
Packaging:
Glass bottle
Catalog number 106040050
also known as 10604
HSN 29222990
Price (INR)
2,770.00
Each
Quantity:
5 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
2,770.00
Each
Chemical Identifiers
CAS103-67-3
IUPAC Namebenzyl(methyl)amine
Molecular FormulaC8H11N
InChI KeyRIWRFSMVIUAEBX-UHFFFAOYSA-N
SMILESCNCC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Refractive index1.5214 to 1.5234 (20°C, 589 nm)
Appearance (Form)Liquid
GC>=96.0 %
Appearance (Color)Clear colorless to light yellow
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only