Bibenzyl, 99%
Bibenzyl, 99%
Bibenzyl, 99%
Acros Organics

Bibenzyl, 99%

CAS: 103-29-7 | C14H14 | 182.27 g/mol
Have Questions?
Catalog NumberQuantityPackaging
106230050
also known as 10623
5 gGlass bottle
Catalog number 106230050
also known as 10623
HSN 29029090
Price (INR)
3,267.00
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
3,267.00
Each
Chemical Identifiers
CAS103-29-7
IUPAC Name(2-phenylethyl)benzene
Molecular FormulaC14H14
InChI KeyQWUWMCYKGHVNAV-UHFFFAOYSA-N
SMILESC(CC1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow
Infrared spectrumConforms
GC>=98.5 %
Appearance (Form)Powder or crystals and/or chunks
Melting point49°C to 53°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only