9-Bromoanthracene, 96%
9-Bromoanthracene, 96%
9-Bromoanthracene, 96%
Acros Organics

9-Bromoanthracene, 96%

CAS: 1564-64-3 | C14H9Br | 257.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 106640050
also known as 10664
HSN 29039990
Price (INR)
6,131.00
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
6,131.00
Each
Chemical Identifiers
CAS1564-64-3
IUPAC Name9-bromoanthracene
Molecular FormulaC14H9Br
InChI KeyZIRVQSRSPDUEOJ-UHFFFAOYSA-N
SMILESBrC1=C2C=CC=CC2=CC2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)Yellow to green to brown
GC>=95.0 %
Appearance (Form)Powder or crystals or needles
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only