2-Bromobutane, 99+%
2-Bromobutane, 99+%
2-Bromobutane, 99+%
Acros Organics

2-Bromobutane, 99+%

CAS: 78-76-2 | C4H9Br | 137.02 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
100 mL
2.5 L
Packaging:
Glass bottle
Catalog number 106785000
also known as 10678
HSN 29033919
Price (INR)
13,280.00
Each
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
13,280.00
Each
Chemical Identifiers
CAS78-76-2
IUPAC Name2-bromobutane
Molecular FormulaC4H9Br
InChI KeyUPSXAPQYNGXVBF-UHFFFAOYNA-N
SMILESCCC(C)Br
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
GC>=99.0 %
Appearance (Color)Clear colorless to yellow-brown
Infrared spectrumConforms
Refractive index1.4359 to 1.4379 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only