Product Image
Thermo Scientific™
HR-ICP-MS System Nebulizers
Choose nebulizers to match your changing analysis requirements from this range compatible with the Thermo Scientific™ ELEMENT™ Series HR-ICP-MS system.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberIncludes
1069400U-5000 AT+ no memb desolv
1069410U-6000 AT+ with memb desolv
1083911PFA Nebulizer (100 μL/Min)
1083921PFA Nebulizer (20 μL/Min)
1110720Glass Nebulizer 1 mL/min
1135450PFA Nebulizer (50 μL/min)
1301820High Pressure PFA Nebulizer
Catalog number 1069400
Price (INR)
-
Includes:
U-5000 AT+ no memb desolv
Choose Thermo Scientific™ HR-ICP-MS System Nebulizers to match your changing analysis requirements, from this wide range compatible with the Thermo Scientific™ ELEMENT™ Series HR-ICP-MS system.
Gain Performance Improvements with Advanced Accessories
Switch to one of these optional nebulizers to fit your laboratorys changing requirements:
  • PFA Microconcentric Nebulizer for acid-resistant inlet system and standard inlet system (100μL/min)
  • PFA Microconcentric Nebulizer for organic matrix inlet system (20μL/min)
  • PFA Microconcentric Nebulizer for organic matrix inlet system (50μL/min)
  • Glass concentric nebulizer
  • MicroMist Nebulizer 200μL/min (best performance with Twinnabar Spray Chamber)
  • ESI™ High pressure PFA ST3 Nebulizer (for use with ESI Apex Q Desolvating Unit)
Specifications
For Use With (Equipment)ELEMENT Series HR-ICP-MS System
IncludesU-5000 AT+ no memb desolv
TypeSystem Nebulizer
Unit SizeEach