1-Bromoheptane, 99%
1-Bromoheptane, 99%
1-Bromoheptane, 99%
Acros Organics

1-Bromoheptane, 99%

CAS: 629-04-9 | C7H15Br | 179.10 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
5 mL
1 L
Packaging:
Glass bottle
Catalog number 107032500
also known as 10703
HSN 29033919
Price (INR)
10,913.00
Each
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
10,913.00
Each
Chemical Identifiers
CAS629-04-9
IUPAC Name1-bromoheptane
Molecular FormulaC7H15Br
InChI KeyLSXKDWGTSHCFPP-UHFFFAOYSA-N
SMILESCCCCCCCBr
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
Refractive index1.4489 to 1.4509 (20°C, 589 nm)
GC>=98.5 %
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only