2-Bromopropane, 99%
2-Bromopropane, 99%
2-Bromopropane, 99%
Acros Organics

2-Bromopropane, 99%

CAS: 75-26-3 | C3H7Br | 122.99 g/mol
Have Questions?
Catalog NumberQuantityPackaging
107320025
also known as 10732
2.5 LGlass bottle
Catalog number 107320025
also known as 10732
HSN 29033920
Price (INR)
34,489.00
Each
Quantity:
2.5 L
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
34,489.00
Each
Chemical Identifiers
CAS75-26-3
IUPAC Name2-bromopropane
Molecular FormulaC3H7Br
InChI KeyNAMYKGVDVNBCFQ-UHFFFAOYSA-N
SMILESCC(C)Br
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=98.5 %
Appearance (Color)Clear colorless to light brown
Refractive index1.4250 to 1.4270 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only