4-Bromopyridine Hydrochloride, 98%
4-Bromopyridine Hydrochloride, 98%
4-Bromopyridine Hydrochloride, 98%
4-Bromopyridine Hydrochloride, 98%
Acros Organics

4-Bromopyridine Hydrochloride, 98%

CAS: 19524-06-2 | C5H4BrN·HCl | 194.46 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number 107420250
also known as 10742
HSN 29333990
Price (INR)
39,366.00
Each
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
39,366.00
Each
Add to cart
Chemical Identifiers
CAS19524-06-2
IUPAC Name4-bromopyridine hydrochloridyl
Molecular FormulaC5H4BrClN
InChI KeyPSPIMDWZDKGZBO-UHFFFAOYSA-N
SMILES[Cl].BrC1=CC=NC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow-pink
Infrared spectrumConforms
Titration with HClO4>=97.5 %
Appearance (Form)Powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only