N-Bromosuccinimide, 99%
N-Bromosuccinimide, 99%
N-Bromosuccinimide, 99%
Acros Organics

N-Bromosuccinimide, 99%

CAS: 128-08-5 | C4H4BrNO2 | 177.99 g/mol
Have Questions?
Catalog NumberQuantityPackaging
107450100
also known as 10745
10 gGlass bottle
Catalog number 107450100
also known as 10745
HSN 29251900
Price (INR)
5,255.00
Each
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS128-08-5
IUPAC Name1-bromopyrrolidine-2,5-dione
Molecular FormulaC4H4BrNO2
InChI KeyPCLIMKBDDGJMGD-UHFFFAOYSA-N
SMILESBrN1C(=O)CCC1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow
Infrared spectrumConforms
Loss on drying=<1 %
Appearance (Form)Crystalline powder
Titration Iodometric98.5 to 101.5 % (on dry substance)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Coupling reagent, C-X bond formation reagent
RUO – Research Use Only