(±)-1,3-Butanediol, 99%, extra pure
(±)-1,3-Butanediol, 99%, extra pure
(±)-1,3-Butanediol, 99%, extra pure
Acros Organics

(±)-1,3-Butanediol, 99%, extra pure

CAS: 107-88-0 | C4H10O2 | 90.12 g/mol
Have Questions?
Catalog NumberQuantityPackaging
107620025
also known as 10762
2.5 LGlass bottle
Catalog number 107620025
also known as 10762
HSN 29053990
Price (INR)
11,031.00
Each
Quantity:
2.5 L
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
11,031.00
Each
Chemical Identifiers
CAS107-88-0
IUPAC Namebutane-1,3-diol
Molecular FormulaC4H10O2
InChI KeyPUPZLCDOIYMWBV-UHFFFAOYNA-N
SMILESCC(O)CCO
View more
SpecificationsSpecification SheetSpecification Sheet
Color scale=<10 APHA
Refractive index1.4390 to 1.4410 (20°C, 589 nm)
Water=<0.5 %
Appearance (Form)Clear liquid
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only