4-tert-Butylcyclohexanone, 99%
4-tert-Butylcyclohexanone, 99%
4-tert-Butylcyclohexanone, 99%
Acros Organics

4-tert-Butylcyclohexanone, 99%

CAS: 98-53-3 | C10H18O | 154.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
500 g
Packaging:
Plastic bottle
Catalog number 107911000
also known as 10791
HSN 29142200
Price (INR)
10,013.00
Each
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (INR)
10,013.00
Each
Chemical Identifiers
CAS98-53-3
IUPAC Name4-tert-butylcyclohexan-1-one
Molecular FormulaC10H18O
InChI KeyYKFKEYKJGVSEIX-UHFFFAOYSA-N
SMILESCC(C)(C)C1CCC(=O)CC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow
Melting point46°C to 51°C
GC>=98.5 %
Infrared spectrumConforms
Appearance (Form)Powder or crystals and/or chunks
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only