(Carbethoxymethyl)triphenylphosphonium bromide, 98%
(Carbethoxymethyl)triphenylphosphonium bromide, 98%
(Carbethoxymethyl)triphenylphosphonium bromide, 98%
Acros Organics

(Carbethoxymethyl)triphenylphosphonium bromide, 98%

CAS: 1530-45-6 | C22H22BrO2P | 429.29 g/mol
Have Questions?
Catalog NumberQuantityPackaging
108300250
also known as 10830
25 gGlass bottle
Catalog number 108300250
also known as 10830
HSN 29319090
Price (INR)
8,072.00
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
8,072.00
Each
Chemical Identifiers
CAS1530-45-6
IUPAC Name(2-ethoxy-2-oxoethyl)triphenylphosphanium bromide
Molecular FormulaC22H22BrO2P
InChI KeyVJVZPTPOYCJFNI-UHFFFAOYSA-M
SMILES[Br-].CCOC(=O)C[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Titration Argentometric>=97.5 %
Appearance (Color)White
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For C-C Bond Formation (Olefination)
RUO – Research Use Only