Methyl succinyl chloride, 97%
Methyl succinyl chloride, 97%
Methyl succinyl chloride, 97%
Acros Organics

Methyl succinyl chloride, 97%

CAS: 1490-25-1 | C5H7ClO3 | 150.56 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Packaging:
Glass bottle
Catalog number 108401000
also known as 10840
HSN 29183010
Price (INR)
39,673.00
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
39,673.00
Each
Chemical Identifiers
CAS1490-25-1
IUPAC Namemethyl 4-chloro-4-oxobutanoate
Molecular FormulaC5H7ClO3
InChI KeySRXOJMOGPYFZKC-UHFFFAOYSA-N
SMILESCOC(=O)CCC(Cl)=O
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=96.0 %
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
Appearance (Form)Liquid
Refractive index1.4390 to 1.4410 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only