4-chlorobenzylamine, 98%
4-chlorobenzylamine, 98%
4-chlorobenzylamine, 98%
Acros Organics

4-chlorobenzylamine, 98%

CAS: 104-86-9 | C7H8ClN | 141.6 g/mol
Have Questions?
Catalog NumberQuantityPackaging
108791000
also known as 10879
100 gGlass bottle
Catalog number 108791000
also known as 10879
HSN 29214990
Price (INR)
34,963.00
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
34,963.00
Each
Chemical Identifiers
CAS104-86-9
IUPAC Name1-(4-chlorophenyl)methanamine
Molecular FormulaC7H8ClN
InChI KeyYMVFJGSXZNNUDW-UHFFFAOYSA-N
SMILESNCC1=CC=C(Cl)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive index1.5576 to 1.5610 (20°C, 589 nm)
GC>=97.5 %
Infrared spectrumConforms
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only