Product Image
Gibco™

Sf-900™ II SFM (1X)

Sf-900™ II SFM is a serum-free, protein-free insect cell culture medium optimized for the growth and maintenance of Spodoptera frugiperdaRead more
Have Questions?
Catalog NumberQuantity
1090215310 x 500 mL
Catalog number 10902153
Price (INR)
-
Quantity:
10 x 500 mL
Customize this product
Sf-900™ II SFM is a serum-free, protein-free insect cell culture medium optimized for the growth and maintenance of Spodoptera frugiperda (Sf9 and Sf21) cells and for large-scale production of recombinant proteins expressed using the baculovirus expression vector system (BEVS). This medium is suitable for suspension and monolayer culture methods and supports growth of other lepidopteran cell lines.

Gibco™ Sf-900™ II SFM features:

• Superior long-term, high density growth
• Optimized for recombinant protein production
• Serum-free, protein-free, ready-to-use formulation
• Scalable in bioreactors

Superior Long-Term, High Density Growth
Spodoptera frugiperda (Sf9) cells grown in Sf-900™ II SFM achieve maximum cell densities of 9 to 12 × 106 cells/mL, a significant improvement over competitor's formulations and Grace’s medium (see Product Manual). Increases in maximum cell densities of 20-100% are also observed with the Lymantria dispar (Gypsy moth) and Trichoplusia ni (Tn-368; Cabbage Looper) cell lines. Gibco™ Sf-900™ II SFM is capable of supporting the cultures to >20 passages.

Optimized for Recombinant Protein Production
Traditionally, Grace's medium supplemented with 10% FBS has been used for recombinant protein expression. Sf-900™ II SFM is an improved serum-free, protein-free medium designed for growth of Sf9 and other lepidopteran cell lines and production of insect virus and rDNA proteins.

Serum-Free, Protein-Free, Ready-to-Use Formulation
Gibco™ Sf-900™ II SFM is a serum-free, protein-free medium that allows for much easier purification of your protein of interest. Sf-900™ II SFM is ready to use; it does not require addition of serum, glutamine or surfactants. Cells adapted to other commercially available serum-free media can be subcultured directly into Sf-900™ II SFM, usually without any further adaptation. Cells usually require some adaptation from serum-containing formulations.

Scalable in Bioreactors
Utility of Sf-900™ II SFM in larger scale cell culture systems was demonstrated with a 5L Celligen™ bioreactor. Successful infections with rAcNPV were carried out producing rβ-Gal and rEPO (see Product Manual).

Product Use
For Research Use Only: Not intended for animal or human diagnostic or therapeutic use. Customers using Gibco™ Sf-900 II SFM in a manufacturing process, who have a submission with the FDA, may request a letter of authorization from us to reference our Type II Drug Master File (DMF).

cGMP Manufacturing and Quality System
Gibco™ Sf-900 II SFM is manufactured at a cGMP compliant facility, located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cell LineSf21, Sf9
Cell TypeInsect Cell
Product LineGibco, Sf-900
Product TypeInsect Cell Serum Free Medium (SFM)
Quantity10 x 500 mL
SpeciesS. frugiperda, Spodoptera frugiperda
ClassificationSerum-free
FormLiquid
Serum LevelSerum-free
With AdditivesGlutamine
Unit SizeEach
Contents & Storage
Storage conditions: 2-8°C. Protect from light
Shipping conditions: Ambient
Shelf life: 12 months from date of manufacture

Frequently asked questions (FAQs)

Can Sf9 cells in Sf-900 III SFM (Cat. No. 12659017) be thawed and grown in Sf-900 II SFM instead?

It should be okay to thaw the cells into Sf-900 II SFM. This is a richer media compared to the Sf-900 III SFM so the cells would have an easy time adapting. We would recommend taking the cells through 3 passages in the new medium before using them for any experiments as that they have enough time to adapt.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

What antimicrobials can be used in insect culture and at what concentration?

Many antibiotics are suitable for use with insect cells. The following antibiotics are commonly used:
- Penicillin/Streptomycine: 50-100 U/mL; 50-100 µg/mL
- Amphotericin B (Fungizone antimycotic): 0.25 µg/mL
- Gentamicin: 0.5 mL of 10 mg/mL solution in 500 mL media (final concentration: 10 µg/mL)

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Is it necessary to heat-inactivate my serum before adding it to my medium?

Heat inactivation is not necessary. Our team has routinely used serum that has not been heat-inactivated, and we have not observed any effect on cell growth or morphology.

Many cells do not require heat-inactivated FBS. Some cells prefer heat-inactivated FBS. For instance, we use heat-inactivated FBS for our insect cell lines, i.e., Sf9 and Sf21 cells.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Do you offer serum-free insect media?

Yes, we offer a variety of serum-free insect media. Please go here (http://www.thermofisher.com/us/en/home/life-science/cell-culture/insect-cell-culture/insect-cell-culture-misc/serum-free-media.html?icid=cvc-insect-media-c2t2) to view the media we offer and the differences between them.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Citations & References (1)

Citations & References
Abstract
Decreased metallation and activity in subsets of mutant superoxide dismutases associated with familial amyotrophic lateral sclerosis.
Authors: Hayward Lawrence J; Rodriguez Jorge A; Kim Ji W; Tiwari Ashutosh; Goto Joy J; Cabelli Diane E; Valentine Joan Selverstone; Brown Robert H Jr;
Journal:J Biol Chem
PubMed ID:11854284
Over 90 different mutations in the gene encoding copper/zinc superoxide dismutase (SOD1) cause approximately 2% of amyotrophic lateral sclerosis (ALS) cases by an unknown mechanism. We engineered 14 different human ALS-related SOD1 mutants and obtained high yields of biologically metallated proteins from an Sf21 insect cell expression system. Both the ... More