Product Image
Thermo Scientific™
HR-ICP-MS Instrument Torch
Increase ion transmission for improved sensitivity with this instrument torch with guard electrode feature, for the Thermo Scientific™ ELEMENT™ Series HR-ICP-MS analyzers.
Have Questions?
Catalog NumberType
1091250Instrument Torch
Catalog number 1091250
Price (INR)
-
Type:
Instrument Torch
Maintain consistently high performance by using only top quality, dedicated consumables such as this Thermo Scientific™ HR-ICP-MS Instrument Torch for the Thermo Scientific™ ELEMENT Series HR-ICP-MS instrument.
The Guard Electrode (GE) feature of this Torch decreases the ion energy spread to increase ion transmission for improved sensitivity. The GE feature is a requirement for Cold Plasma measurements.
Specifications
For Use With (Equipment)ELEMENT Series HR-ICP-MS System
TypeInstrument Torch
Unit SizeEach