Use the Thermo Scientific™ DRI™ Acetaminophen Calibrator Set with the Thermo Scientific Acetaminophen Serum Toxicology Reagent Kit.
Have Questions?
Catalog Number
Quantity
1091TS1
also known as 1091
1 x 5 mL, 5 x 2 mL
Catalog number 1091TS1
also known as 1091
Price (INR)
-
Quantity:
1 x 5 mL, 5 x 2 mL
Use the Thermo Scientific™ DRI™ Acetaminophen Serum Toxicology Calibrator Set with the Thermo Scientific Acetaminophen Serum Toxicology Reagent Kit.
The DRI Acetaminophen Calibrator Kit contains 5mL of negative calibrator and 2mL each of 10µg/mL, 25µg/mL, 50µg/mL, 100µg/mL and 200µg/mL acetaminophen calibrators in Tris buffer with sodium azide as a preservative.