4-chloroquinoline, 99%
4-chloroquinoline, 99%
4-chloroquinoline, 99%
Acros Organics

4-chloroquinoline, 99%

CAS: 611-35-8 | C9H6ClN | 163.60 g/mol
Have Questions?
Catalog NumberQuantity
110050050
also known as 11005
5 g
Catalog number 110050050
also known as 11005
HSN 29334900
Price (INR)
48,669.00
Each
Quantity:
5 g
Request bulk or custom format
Price (INR)
48,669.00
Each
Chemical Identifiers
CAS611-35-8
IUPAC Name4-chloroquinoline
Molecular FormulaC9H6ClN
InChI KeyKNDOFJFSHZCKGT-UHFFFAOYSA-N
SMILESClC1=C2C=CC=CC2=NC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Low melting solid
GC>=98.5 %
Appearance (Color)Light yellow
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only