Cholesteryl pelargonate, +99%, Thermo Scientific Chemicals
Cholesteryl pelargonate, +99%, Thermo Scientific Chemicals
Cholesteryl pelargonate, +99%, Thermo Scientific Chemicals
Acros Organics

Cholesteryl pelargonate, +99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Glass bottle
Catalog number 110271000
also known as 11027
HSN 29159090
Price (INR)
20,310.00
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
20,310.00
Each
Chemical Identifiers
CAS1182-66-7
IUPAC Name9a,11a-dimethyl-1-(6-methylheptan-2-yl)-1H,2H,3H,3aH,3bH,4H,6H,7H,8H,9H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-7-yl nonanoate
Molecular FormulaC36H62O2
InChI KeyWCLNGBQPTVENHV-UHFFFAOYNA-N
SMILESCCCCCCCCC(=O)OC1CCC2(C)C3CCC4(C)C(CCC4C3CC=C2C1)C(C)CCCC(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Specific optical rotation-28° to -32° (20°C, 589 nm) (C=2, CHCl3)
TLC>=99.0 %
Solubilityalmost colorless
Appearance (Color)White to almost white
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only