Dynabeads™ Pan Mouse IgG
Invitrogen™

Dynabeads™ Pan Mouse IgG

Dynabeads Pan Mouse IgG binds monoclonal mouse IgGs of your choice for specific cell isolation or depletion.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
11040D2 mL
110415 mL
110425 x 5 mL
Catalog number 11040D
Price (INR)
25,851.00
Online offer
Ends: 31-Mar-2026
43,084.00
Save 17,233.00 (40%)
Each
Quantity:
2 mL
Price (INR)
25,851.00
Online offer
Ends: 31-Mar-2026
43,084.00
Save 17,233.00 (40%)
Each

Dynabeads Pan Mouse IgG binds monoclonal mouse IgGs of your choice for specific cell isolation or depletion. It will bind all subclasses of mouse IgG and is Fc specific. Low amount of IgG is required for binding. All cells from all species except from mouse can be isolated with this product, depending only on the specific antibody of choice.

  • Depletion—cells can be depleted from a sample
  • Positive isolation—cells can be positively isolated from a sample for molecular applications
  • Negative isolation—cells can be negatively isolated from a PBMC sample by adding an antibody cocktail of mouse IgGs towards the unwanted cells. Compatible with flow cytometry.

Note:

For cellular assays we recommend positive isolation followed by release of beads from the cells. Recommended product for bead-free cells: Dynal CELLection Pan Mouse IgG (Cat. No. 115-31D) in combination with your mouse IgG.

Starting samples:

Whole blood, PBMC, buffy coat, tissue digests.

Dynabeads Pan Mouse IgG binds monoclonal mouse IgGs of your choice for specific cell isolation or depletion. It will bind all subclasses of mouse IgG and is Fc specific. Low amount of IgG is required for binding. All cells from all species except from mouse can be isolated with this product, depending only on the specific antibody of choice.

  • Depletion - cells can be depleted from a sample
  • Positive isolation - cells can be positively isolated from a sample for molecular applications
  • Negative isolation - cells can be negatively isolated from a PBMC sample by adding an antibody cocktail of mouse IgGs towards the unwanted cells. Compatible with flow cytometry.

Note:

For cellular assays we recommend positive isolation followed by the release of beads from the cells. Recommended product for bead-free cells: CELLection Pan Mouse IgG (Cat. No. 115-31D) in combination with your mouse IgG.

Starting samples:

Whole blood, PBMC, buffy coat, tissue digests.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Antibody FragmentWhole antibody
Cell TypeAll cells from all species, except mouse
No. of CellsProcesses ∼8x 10^8 cells total
Output Viability>95%
Product LineDynabeads
Purity or Quality GradeResearch Grade
Quantity2 mL
Sample TypePBMC, Tissue Digests, Blood
Shelf Life36 Months (from date of manufacture)
Shipping ConditionRoom Temperature
Starting Material Cell No.1 x 10^7 PBMCs per isolation
Target SpeciesAll species, except mouse
ColorBrown
Diameter (Metric)4.5 μm
For Use With (Application)Cell Depletion, Negative Isolation, Positive Isolation
MaterialMagnetic Polystyrene
Product TypeAntibody Coated Bead
Unit SizeEach
Contents & Storage
This Product Contains: 2 mL Dynabeads™ Pan Mouse IgG coated with monoclonal human anti-mouse IgG. The monoclonal antibody is a unique Fc-specific human anti-mouse IgG4, and it shown no cross reactivity to human, rat, hamster, goat, rabbit and guinea pig IgG.
Store at 2°C to 8°C.